Cell culture microplates, cell-repellent surface, CELLSTAR®
Supplier: Greiner Bio-One
651970_6
GREI651970_6EA 0 SGD
GREI651970_6
Cell culture microplates, cell-repellent surface, CELLSTAR®
Microplates Cell Culture Plates
These sterile, PS multiwell cell culture plates have a cell-repellent surface to effectively inhibit cell attachment. Achieved through a chemical polymer modification, the surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
  • Effectively inhibits cell adhesion
  • Sterile
  • Lid with condensation rings
  • Other formats available (flasks, dishes, multiwell plates)
Characterised by low cell attachment, the cell-repellent surface is ideal for applications such as spheroid culturesMore Product Information