Lids for 96-well microplates
Supplier: Greiner Bio-One
656101_100
GREI656101_100EA 0 SGD
GREI656101_100
Lids for 96-well microplates
Lids Microplate Lids
These high profile lids, height 9 mm, are designed to protect against contamination and evaporation during sample storage and cell culture. Lids without condensation rings are mainly used with non tissue culture treated microplates. Lids with condensation rings are recommended for cell culture applications, since they inhibit cross contamination.