Cell culture multiwell plates, CELLSTAR®
Supplier: Greiner Bio-One
662160_100 662102_100 657185_100
GREI662160_100EA 0 SGD
GREI662160_100 GREI662102_100 GREI657185_100
Cell culture multiwell plates, CELLSTAR®
Microplates Cell Culture Plates
PS, with lid, sterile. Cell culture multiwell plates are available either with a physically treated surface (for adherent cell cultures) or with a hydrophobic surface (for suspension cultures and hybridoma cells). All plates are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • High clarity and low autofluorescence
  • Lid enables optimal gas exchange with the lowest possible evaporation
  • Single position lids prevent cross contamination
  • Alphanumeric well labelling
  • Compatible with automated systems
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 (241002); ISO 50001 (241002)