Cell culture inserts for multiwell plates, ThinCert™
Supplier: Greiner Bio-One
662638_48 665630_48
GREI662638_48EA 0 SGD
GREI662638_48 GREI665630_48
Cell culture inserts for multiwell plates, ThinCert™
Microplates Insert Plates and Inserts
Clear PS housing with PET capillary pore membranes, cell culture surface treated, sterile. Combining six different membrane specifications (pore size and density) in geometries to fit 6,12 and 24 well plates, the ThinCert™ cell culture inserts are suitable for a wide range of applications including transportation, secretion and diffusion studies, migrational experiments, cytotoxicity testing, co cultures, Trans Epithelial Electric Resistance (TEER) measurements, as well as primary cell culture.
  • Physical surface treatment optimises cell adherence and growth
  • Hanging geometry ensures
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable