Glass bottom microplates, SensoPlate™
Supplier: Greiner Bio-One
662892_12
GREI662892_12EA 1280 SGD
GREI662892_12
Glass bottom microplates, SensoPlate™
Microplates HTS Microplates
Optically clear borosilicate glass bottom (light path 175±15 µm) and black PS frame.
  • All well formats available: 24, 96, 384 and 1536 wells
  • Standard SBS footprint
  • SensoPlate™ Plus option for high throughput screening applications
  • Free of DNase, RNase and human DNA, and non-pyrogenic
SensoPlate™ glass bottom microplates are designed for fluorescence correlation spectroscopy. The glass bottom allows transmission measurements in the wavelength range above 350 nm. Microscopic applications, such as confocal microscopy, are another potential area ofMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 (241002); ISO 50001 (241002)