Cell culture insert plates, ThinCert™
Supplier: Greiner Bio-One
665110_60
GREI665110_60EA 2130 SGD
GREI665110_60
Cell culture insert plates, ThinCert™
Microplates Insert Plates and Inserts
PS, sterile, with lid. Designed for use in air-lift culture where tissue is cultivated in permeable membrane supports (cell culture inserts), with the cell culture medium nourishing the cells from below the membrane and the air exerting its influence from above. If cell culture inserts and conventional cell culture plates are utilised in the air-lift culture, the available volume of culture medium is severely limited. This reduced volume results in an elevated nutrient consumption rate from the lower compartment and an increased number of medium changes.
  • Optimised for use with ThinCert™
More Product Information