PCR tubes, with flat or domed cap, Sapphire
Supplier: Greiner Bio-One
682201_1000
GREI682201_1000EA 0 SGD
GREI682201_1000
PCR tubes, with flat or domed cap, Sapphire
Tubes PCR Tubes
Manufactured out of medical-grade polypropylene without interfering additives.
  • Thin-walled for efficient heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Suitable for real-time / quantitative PCR
  • Comprehensive thermal cycler compatibility
Polypropylene tubes feature thin-wall construction for optimal heat transfer from the thermocycler block to the reaction mixture.