Serological pipettes
Supplier: Greiner Bio-One
606180_200 607180_200 768160_100 607160_200 760160_200
GREI606180_200EA 0 SGD
GREI606180_200 GREI607180_200 GREI768160_100 GREI607160_200 GREI760160_200
Serological pipettes
Pipettes Serological Pipettes
High-quality transparent PS, graduated, sterile.
  • Clear and easy-to-read graduation, with a vertical coloured stripe on the 1, 2, 5, 10 and 25-ml pipettes
  • Increased capacity thanks to negative graduation
  • Cotton insert to stop liquids from rising up into the pipetting device
  • Colour code conforms to international standards to facilitate pipette selection
  • Free from DNase, RNase and human DNA, non pyrogenic and non cytotoxic
Packaging: Bulk packed in plastic resealable zip sachets, individually wrapped in peelable plastic/paper, or individually packaged inMore Product Information