Strip plates, 96-well, for ELISA
Supplier: Greiner Bio-One
762061_400 762001_400
GREI762061_400EA 260 SGD
GREI762061_400 GREI762001_400
Strip plates, 96-well, for ELISA
Microplates Immunology Microplates and Strips
PS, without lid. Microplates in strip format offer the advantage of greater flexibility. Individual strips can be removed from the support frame so that the number of tests to be performed can be adjusted to the number of samples, and is not predetermined by the microplate format used. Strip plates are available with high binding surfaces that display a higher protein binding capacity for polar proteins (MICROLON® 600), medium binding surfaces, which are more hydrophobic and are therefore better suited for non-polar proteins and peptides (MICROLON® 200), black (FLUOTRAC™) or white (LUMITRAC™)More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Environmental certification ISO 14001 (241002); ISO 50001 (241002) ISO 14001 (241002); ISO 50001 (241002)