Cell culture plates, 384-well, CELLSTAR®
Supplier: Greiner Bio-One
781080_32
GREI781080_32EA 340 SGD
GREI781080_32
Cell culture plates, 384-well, CELLSTAR®
Microplates Cell Culture Plates
PS, cell culture-treated, sterile. All plates are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • F-bottom wells with excellent optical properties are ideal for microscopic applications
  • Clear bottom plates have pigmented walls and a transparent thin film bottom, ideal of fluorescence or luminescence detection methods
  • Lid enables gas exchange with minimal evaporation
Colour: Solid bottom plates are available clear, black or white; µClear® (clear) bottom plates are available black or white
Colour: Solid bottomMore Product Information