Biohazard spill kit
Supplier: GUEST
H8616
GUESH8616EA 220 SGD
GUESH8616
Biohazard spill kit
Spill Control Materials
The biohazard spill kit contains everything needed to clean up spills of blood or other biohazardous fluid quickly and safely. Each kit contains.
  • Haz-Tab granules, which are formulated using sodium dichloroisocyanurate, a powerful chlorinating agent effective against bacteria, fungi and viruses; poured onto the spill they absorb and disinfect simultaneously
  • A scoop and scraper to transfer the spill, including broken glass, to the yellow incineration bag for disposal
  • Tablets to enable preparation of a 10000 ppm solution of chlorinating agent, with which
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable