Phosphate standard solutions
Supplier: Hach
1436832
HACH1436832EA 74.9 SGD
HACH1436832
Phosphate standard solutions
Standards Environmental Analysis Standards
Standard solution for accuracy checks in phosphate (phosphorus) determinations.