Hardness reagent, UniVer® 3
Supplier: Hach
21320H
HACH21320HEA 97.3 SGD
HACH21320H
Hardness reagent, UniVer® 3
Environmental Test Kits
This combined indicator and buffer reagent is used for determination of total hardness by titration methods.