Sulphuric acid 0.0175 mol/l (0.035 N) in aqueous solution

Supplier: Hach

2349732
HACH2349732EA 59.8 SGD
HACH2349732
Sulphuric acid 0.0175 mol/l (0.035 N) in aqueous solution
Sulphuric acid
Formula: H₂SO₄
Density: 1,01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR