Hexamethylenetetramine

Supplier: Hach
Warning

2603999
HACH2603999EA 324 SGD
HACH2603999
Hexamethylenetetramine
Methenamine
Formula: C₆H₁₂N₄
Melting Pt: 280 °C
Density: 1,33 g/cm³ (20 °C)
Flash Pt: 250 °C
Storage Temperature: Ambient
MDL Number: MFCD00006895
CAS Number: 100-97-0
EINECS: 202-905-8
UN: 1328
ADR: 4.1,III
Merck Index: 13,05994
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3ecbb5df5394322c2150525ce6e9f48b5aadde7&vid=04b623f7f333c7cf159016619cf91fc792af9a3f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR