THM reagent 3
Supplier: Hach
2754142
HACH2754142EA 524 SGD
HACH2754142
THM reagent 3
Environmental Test Kits
Screening tool to optimise treatment processes, to locate areas of high THM levels in distribution systems and to determine levels of trihalomethanes in drinking water samples.
  • For determination of Total Trihalomethanes by the THM Plus™ Method
  • Hach Method 10132
Caution: Corrosive
Order Now
SPECIFICATIONS
Environmentally Preferable