pH buffer solutions, colour coded

Supplier: Hach

2947600
HACH2947600EA 149 SGD
HACH2947600
pH buffer solutions, colour coded
Buffers pH Calibration Buffers

For calibration of pH meters


  • Colour coded for easy identification
  • Accuracy (pH): ±0,02 at 25 °C


Certifications: Traceable to NIST.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR