Conductivity standards, KCL
Supplier: Hach
C20C250
HACHC20C250EA 245 SGD
HACHC20C250
Conductivity standards, KCL
Standards Conductivity Standards
The potassium chloride standard solutions are used for calibrating conductivity cells.