Reference electrodes, Radiometer Analytical
Supplier: Hach
E21M002
HACHE21M002EA 714 SGD
HACHE21M002
Reference electrodes, Radiometer Analytical
Electrodes ISE Electrodes
Radiometer Analytical reference electrodes are available with a variety of reference systems and liquid junctions.
  • Liquid junction: porous pin
  • Red-Rod technology provides a reproducible, fast and accurate response
  • Calomel reference electrode can be used for special purposes
* These electrodes contain mercury, please dispose according to regulations.
** Screw cap electrodes require an external connection cable for connection to a meter.