Reagents for DR 2800, DR 3800 DR 3900, DR 5000 and DR 6000
Supplier: Hach
HPT430
HACHHPT430EA 270 SGD
HACHHPT430
Reagents for DR 2800, DR 3800 DR 3900, DR 5000 and DR 6000
Environmental Test Kits
Tests for all water analysis applications. The cuvette is a closed sytem which is safe for the user and the environment.
  • Accurate and precise measured values
  • Fast and simple procedures
  • Ready-to-use reagent
  • Cuvette tests are approved for consent limits testing