Bench top turbidity meters, TL23
Supplier: Hach
LPV4449900120
HACHLPV4449900120EA 14750 SGD
HACHLPV4449900120
Bench top turbidity meters, TL23
Turbidity Meters
Tungsten or LED light sources, as well as proven optics, make these instruments more reliable to deliver accurate and stable measurements.
  • Improved and intuitive design with guided calibration and verification
  • USB port for data export and sample identification
  • Ratio technology that helps to overcome interference from colour and large particles
  • Coloured tactile LCD with commands
  • 90° detector
Designed for simple use in demanding applications of pharmaceutical, cosmetic, food, chemical and petrochemical industries.
Certifications: US EPA 180.1 or ISO 7More Product Information