ORP combination electrodes, IntelliCAL™ MTC101 / MTC301
Supplier: Hach
MTC10105
HACHMTC10105EA 2650 SGD
HACHMTC10105
ORP combination electrodes, IntelliCAL™ MTC101 / MTC301
Electrodes Redox Electrodes
Digital combination ORP/Redox probes with built-in temperature sensor. Ideal for measuring absolute mV values in wastewater, drinking water and general applications.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Traceability in measurement history: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • User alerted when re-calibration is needed
  • Plug & Play: Can be moved between meters without the need to re-calibrate or re-enter measurement settings
  • Rugged digital
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable