Biomedical freezers –25 to –10 °C
Supplier: HAIER MEDICAL LABORATORY PRO
DW-25L92
HAIEDW-25L92EA 1040 SGD
HAIEDW-25L92
Biomedical freezers –25 to –10 °C
Freezers Laboratory / General Purpose Freezers
Upright biomedical freezers, with manual defrost.
  • Reliable performance: High density foam insulation for rigidity and stable storage temperature
  • LED digital display for clear observation
  • Two alarm functions: High/low temperature, senor error
  • Adjustable temperature: Adjustable freezer compartment range: –25 to –10 °C