Container for Hazardous Waste
Supplier: HAMILTON ROBOTICS
281520 199201 185319
HAMR281520EA 95 SGD
HAMR281520 HAMR199201 HAMR185319
Container for Hazardous Waste
Boxes Waste Disposal Boxes
The box is used with waste chute 736-0847 and 736-0846.
  • Container complete with yellow PE bag, thickness of foil 120 µm
  • Two types of bags for container available
  • Bags are highly resistant to tearing and puncturing
Order Now
SPECIFICATIONS
Environmentally Preferable