Robotic tips (CO-RE Tips) and consumables for Microlab
Supplier: HAMILTON ROBOTICS
235900 235902 235904 235966 235447 235444 235449 235451 235971 235974 235978 235979 235983 235987 235695 235993 235989 235901 235903 235905 235936 235940 235947 235948 235949 235935 235646 235647 235648 235452 235677 235678 235694 184021 184023 194050 194053 235806 235541 235688
HAMR235900EA 1130 SGD
HAMR235900 HAMR235902 HAMR235904 HAMR235966 HAMR235447 HAMR235444 HAMR235449 HAMR235451 HAMR235971 HAMR235974 HAMR235978 HAMR235979 HAMR235983 HAMR235987 HAMR235695 HAMR235993 HAMR235989 HAMR235901 HAMR235903 HAMR235905 HAMR235936 HAMR235940 HAMR235947 HAMR235948 HAMR235949 HAMR235935 HAMR235646 HAMR235647 HAMR235648 HAMR235452 HAMR235677 HAMR235678 HAMR235694 HAMR184021 HAMR184023 HAMR194050 HAMR194053 HAMR235806 HAMR235541 HAMR235688
Robotic tips (CO-RE Tips) and consumables for Microlab
Pipette Tips
Hamilton CO-RE tips were designed for MICROLAB pipetting workstations in order to solve the problem of sealing and alignment plagued by press-fit tip attachment techniques. Each CO-RE disposable tip has a circular groove inside the attachment orifice. An o-ring on the pipetting channel is expanded to fit this groove precisely providing an air-tight seal and straight alignment between tip and pipetting channel.
  • Pressure-based Liquid Level Detection (pLLD) and capacitive LLD (cLLD) with conductive (black) version
  • DNase-/RNase-free, pyrogen-free (endotoxin-
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable