Digital thermometer, Checkfridge®
Supplier: Hanna
HI147-00
HANNHI147-00EA 262 SGD
HANNHI147-00
Digital thermometer, Checkfridge®
Thermometers Digital Thermometers
The Checkfridge® is a handy and precise thermometer adapted to accurate and reliable temperature monitoring. It allows continuous measurements in cold rooms, freezers and refrigeration units. It is possible to monitor inside temperature without having to open the door.
  • Magnetic backing for adhesion to metallic surfaces
  • Test switch conveniently located on the face of the thermometer
  • Unique Cal-Check™ mode feature performs an internal calibration check in only a few seconds
Stainless steel probe (Lר: 40×5 mm) and one-meter cable.
Cal-Check™ function.More Product Information