Test sieves
Supplier: NEXOPART
206158772
HAVE206158772EA 664 SGD
HAVE206158772
Test sieves
Sieves Test Sieves
These ALPINE air jet test sieves are suitable for fully automatic sieve recognition. These test sieves are made of brass/stainless steel frame with metal wire cloth made from stainless steel.
Certifications: According to EN ISO/IEC 17025, DIN EN 10204.
Delivery information: Supplied with serial number, inspection certificate and calibration certificate on request.