Counting chamber, Modified Fuchs-Rosenthal
Supplier: HAWKSLEY AND SONS
AC6000
HAWKAC6000EA 496 SGD
HAWKAC6000
Counting chamber, Modified Fuchs-Rosenthal
Slides Counting Chambers
This two cell counting chamber has metallised platforms.
  • Total area of ruling: 1/6 mm²
  • Rhodium coating
Delivery information: Supplied with two cover glasses.
Order Now
SPECIFICATIONS
Environmentally Preferable