Portable centrifuge kits
Supplier: HEATHROW
120568 120569 120570
HEAT120568EA 1250 SGD
HEAT120568 HEAT120569 HEAT120570
Portable centrifuge kits
Centrifuges Microcentrifuges
The Portable centrifuge kit by Heathrow Scientific offers a convenient way to spin down and prepare samples outside of a conventional laboratory environment.
  • Kit enables immediate on-site testing for faster results
  • Includes five year warranty
The centrifuge kit enables the processing of samples before deterioration can take place and can be used in remote environments where a normal electrical supply is not an option.
Conveniently carry the centrifuge and its components securely in the DuraPorter® transport box that has a specially designed insertMore Product Information