Combined centrifuge and magnetic stirrer, MagFuge®
Supplier: HEATHROW
120581 120582
HEAT120581EA 2740 SGD
HEAT120581 HEAT120582
Combined centrifuge and magnetic stirrer, MagFuge®
Centrifuges Microcentrifuges
Convenient high-speed centrifuge and magnetic stirrer combined into one easy to use unit with a digital display, one button programming and tool free rotor exchange.
  • An easy to use back-lit digital control panel displays time, speed and operation mode
  • Manual lid override allows users to open the lid in case of a power failure and prevents loss of samples
  • Safety features ensure it will not operate when the lid is open, a rotor is jammed, or the unit is out of balance
  • Can be used at 2 to 40 °C max. Relative Humidity Non-condensing, 80% for temperatures up to
More Product Information