Mini centrifuges, Sprout® plus
Supplier: HEATHROW
120611 120610
HEAT120611EA 918 SGD
HEAT120611 HEAT120610
Mini centrifuges, Sprout® plus
Centrifuges Microcentrifuges
Mini centrifuge with ergonomic design, easy to operate and gives increased efficiency.
  • Small footprint and quiet operation
  • ABS plastic housing and polycarbonate lid enable the unit to be easily cleaned
  • Palm-shaped lid, suitable for left or right handed users allows quick extraction of tubes
  • Weighted rubberized base eliminates potential movement and further dampens vibration and noise
  • Suitable for worldwide voltages 100 to 240 V, supplied with four different plug adapters
  • Lid locks for added protection when the unit is in use, larger lid accommodates longer
More Product Information