High speed mini-centrifuge, GUSTO®
Supplier: HEATHROW
HS10050
HEATHS10050EA 2680 SGD
HEATHS10050
High speed mini-centrifuge, GUSTO®
Centrifuges Microcentrifuges
The Gusto® is a high speed, space saving mini centrifuge. This compact centrifuge is ideal for protocols calling for fast spins with a higher RCF requirement.
  • Tool-free rotor exchange
  • Adjustable time and speed
  • Durable ABS body and polycarbonate lid
  • Powered by 12V DC
  • Push button lid release
  • Manual override allows user to open lid in case of power failure
  • Long-life maintenance-free motor
  • Easy to clean design
The Gusto® is ideal for PCR®, microfilter cell separation, clinical and diagnostic applications, and HPLC protocols. The micro-centrifuge has aMore Product Information