Biohazard Bags
Supplier: HEATHROW
HS10320 HS10321 HS10322 HS10323
HEATHS10320EA 309 SGD
HEATHS10320 HEATHS10321 HEATHS10322 HEATHS10323
Biohazard Bags
Bags Waste Disposal Bags
These biohazard waste bags are for personal or general lab use, sizes to suit all needs.
  • Includes an indicator patch that darkens when autoclaved
  • Autoclavable to 121 °C (250 °F)
  • Disposable
A range of biohazard collection bags printed with biohazard warning symbol and precautions in English, Spanish, and French.
Ordering information: Epoxy coated steel wire holder available for bags HEATHS10320.
Order Now
SPECIFICATIONS
Environmentally Preferable