Bottles and tube racks
Supplier: HEATHROW
HS120085 HS120086
HEATHS120085EA 60.5 SGD
HEATHS120085 HEATHS120086
Bottles and tube racks
Racks Bottle Racks
Wire, epoxy coated.
  • Rack suitable for Bijou bottles and low profile tubes up to 25 mm diameter
  • Mesh bottom support
  • Chemically-resistant steel wire racks
For transporting and handling samples.
Order Now
SPECIFICATIONS
Environmentally Preferable