Nylon microtube rack
Supplier: HEATHROW
HS120241
HEATHS120241EA 199 SGD
HEATHS120241
Nylon microtube rack
Racks Tube Racks
The 24-Well Microtube rack is made of reinforced nylon, and offers superior chemical and heat resistance.
All racks are rated for use in temperatures ranging from –40 to +140 °C (–40 to +284 °F).
Integrated handles enable easy and safe transportation. Racks are stackable and available in two formats for small or large requirements. Autoclavable.