Wash bottles, safety labelled and self venting
Supplier: HEATHROW
HS120250 HS120251 HS120252 HS120253 HS120255 HS120256 HS120257
HEATHS120250EA 157 SGD
HEATHS120250 HEATHS120251 HEATHS120252 HEATHS120253 HEATHS120255 HEATHS120256 HEATHS120257
Wash bottles, safety labelled and self venting
Bottles Wash Bottles
Clear and informative safety information.
  • Wide bottle mouth makes filling easier
  • Non-metallic vent prevents the risk of solvent leakage
  • Bottle is made from LDPE, with a HDPE closure
The Wash Bottle is specific to the product contained and includes the following: the Chemical Abstract Service (CAS) No., National Fire Protection Association (NFPA) codes and symbols, Protective Equipment Symbol (PES), Target Organ Information, and Hazard Codes. There is also the option of using other liquids within the bottles and creating the appropriate safetyMore Product Information