Blood collection tray
Supplier: HEATHROW
HS120261
HEATHS120261EA 316 SGD
HEATHS120261
Blood collection tray
Trays/Dishes/Pots Blood Collection Trays
Organise your phlebotomy, I.V. start or catheter insertion supplies with this spacious, robust and easy to clean carry tray.
  • Lightweight easy-to-clean ABS construction
  • Comfort grip handle tested to hold up to 10 kg
Customize the compartments to suit your personal needs with the dividers provided.
Delivery information: Includes a 17 mm well, 60 place tube rack.