Colony counter, eCount™
Supplier: HEATHROW
HS120000 HS15092ZP
HEATHS120000EA 695 SGD
HEATHS120000 HEATHS15092ZP
Colony counter, eCount™
Colony Counters
The all-in-one eCount™ combines a multifunctional electronic counter and a Sharpie® pen for marking Petri dishes to prevent missing or double-counting colonies.
  • Five-digit display
  • Touch controls for multiple counts: Up to 32 different zones
  • Counter automatically beeps with each count, or select the LED flash mode for a visual alert of each count
  • Working temperature from 0 to 50 °C
  • Light ergonomic body balanced for easy manipulation
Designed for rapid, accurate counts. Press the lock key to write with the Sharpie® pen without counting. The Sharpie® penMore Product Information