Clamp holders for pipettes, Clamp-ette®
Supplier: HEATHROW
HS20162A
HEATHS20162AEA 80.3 SGD
HEATHS20162A
Clamp holders for pipettes, Clamp-ette®
Pipette Stands
PP, coloured.
  • Stable, sturdy construction
  • For one to three pipettes of the most widely used brands
  • Secures to tabletops, benches and shelves with a twist of the fingertips
  • Autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable