Storage and organisation system
Supplier: HEATHROW
HS20612
HEATHS20612EA 192 SGD
HEATHS20612
Storage and organisation system
Pipette Stands
ABS, coloured. For standard and tissue culture pipettes, and pipette boxes. Strong, stable construction.
  • Suitable for standard and tissue culture pipettes from 0,1 to 25 ml
  • Four compartments with side viewing windows allow a clear view, even when several systems are arranged next to each other
  • Lateral clamp holders for pipetting aids or pipetting bulbs