Racks for microcentrifuge tubes
Supplier: HEATHROW
HS2166
HEATHS2166EA 41.6 SGD
HEATHS2166
Racks for microcentrifuge tubes
Racks Tube Racks
Polyfoam.
  • For a variety of tubes
  • Floating rack
  • Beaker-sized rack comes with an optional handle