Petri dish dispenser
Supplier: HEATHROW
HS23452
HEATHS23452EA 193 SGD
HEATHS23452
Petri dish dispenser
Racks Petri Dish Racks
Acrylic. This sturdy dispenser will hold up to twenty Ø 90 mm Petri dishes in two easy to access partitions.
  • Mounting screws included
  • On the counter or on a wall
  • Solid and clear design conception