Storage units, MagLab™ series
Supplier: HEATHROW
HS23500 HS23502 HS23503 HS23504
HEATHS23500EA 220 SGD
HEATHS23500 HEATHS23502 HEATHS23503 HEATHS23504
Storage units, MagLab™ series
Boxes Storage Boxes
The MagLab™ series allows common laboratory products to be organised off the benchtop.
ABS, coloured.
  • MagRack™ holds four 50 ml tubes and nine 15 ml tubes
  • MagWipe™ holds a standard (11×21 cm) Kimwipes® box
  • MagPette™ holds two pipettors (fits most popular styles)
  • MagBox™ holds miscellaneous accessories
Available individually or as a set of four, each unit is secured by magnets that hold onto most metal surfaces.