Cell spreaders, Delta™
Supplier: HEATHROW
HS8151
HEATHS8151EA 284 SGD
HEATHS8151
Cell spreaders, Delta™
Spreaders Cell Spreaders
Triangular shaped spreading bar has an edgeless design which prevents tearing of the agar and head shape that distributes pressure evenly for smooth spreading. Handle flexes slightly under pressure and is rippled for easy manipulation. Dimensions 172×37×4 mm.