Ring weights for vessels
Supplier: HEATHROW
HS8881A HS8881B HS8881C HS8881D HS8882A HS8882B HS8882D
HEATHS8881AEA 75.4 SGD
HEATHS8881A HEATHS8881B HEATHS8881C HEATHS8881D HEATHS8882A HEATHS8882B HEATHS8882D
Ring weights for vessels
Weights Flask Weights
Lead core, with blue vinyl coating. Open or closed design. Open design is also suitable for vessels with lateral connections. For stabilising glass or plastic vessels on shakers or in water and oil baths, etc. The weights prevent tipping. The coating prevents the vessels from being scratched or causing damage to other surfaces.
  • Good chemical resistance
  • High temperature resistance
  • Available in various sizes for use with a range of vessel sizes
Order Now
SPECIFICATIONS
Environmentally Preferable