Counting chambers, Fuchs-Rosenthal
Supplier: GLASWARENFABRIK KARL HECHT
40449702 40449703
HECH40449702EA 85.1 SGD
HECH40449702 HECH40449703
Counting chambers, Fuchs-Rosenthal
Slides Counting Chambers
This ruling is used for counting leukocytes, eosinophils and cells in cerebro-spinal fluid.
  • Total area of ruling: 16 mm²
  • Subdivision: 16 square groups of 1 mm² each
  • 16 mini squares (0,25×0,25 mm) each
Order Now
SPECIFICATIONS
Environmentally Preferable