Centrifuge tubes, cylindrical, round bottom
Supplier: GLASWARENFABRIK KARL HECHT
40950001
HECH40950001EA 49.1 SGD
HECH40950001
Centrifuge tubes, cylindrical, round bottom
Tubes Centrifuge Tubes
AR-Glas® (soda-lime glass).
  • Ungraduated or graduated
  • Good chemical resistance
Round bottom, rimless.