Centrifuge tubes, cylindrical, round bottom
Supplier: GLASWARENFABRIK KARL HECHT
40958016
HECH40958016EA 122 SGD
HECH40958016
Centrifuge tubes, cylindrical, round bottom
Tubes Centrifuge Tubes
Borosilicate glass 3.3.
  • Ungraduated or graduated
  • Excellent chemical resistance
  • Excellent temperature resistance
Round bottom, rimless