Cavity slides for immunofluorescence
Supplier: GLASWARENFABRIK KARL HECHT
42424010 42423010 42420010 42422010 42421010
HECH42424010EA 236 SGD
HECH42424010 HECH42423010 HECH42420010 HECH42422010 HECH42421010
Cavity slides for immunofluorescence
Slides Cavity Slides
Black (or blue) epoxy-coated slides.
Specifically designed for immunofluorescence.
Order Now
SPECIFICATIONS
Environmentally Preferable