Staining jars
Supplier: GLASWARENFABRIK KARL HECHT
42480010 42481010 42482010
HECH42480010EA 56.7 SGD
HECH42480010 HECH42481010 HECH42482010
Staining jars
Jars Staining Jars
Soda-lime glass.
  • For 10 standard slides  (76×26 mm)
Staining system contains staining jar, rack and handle made from stainless steel.
Ordering information: Insert and handle must be ordered separately.
Order Now
SPECIFICATIONS
Environmentally Preferable